COM448618188 Combi-Blocks, Inc.

2-Chloro-5-methoxy-4-methylbenzeneboronic acid

  • MDL number MFCD11870203
  • Cas Number : 2096335-81-6
  • Molecular Weight : 200.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2184
Availability details for
Quantity
  • Availability
  • Add to Cart