COM448618192 Combi-Blocks, Inc.

5-Cyano-1-methyl-1H-pyrrol-2-ylboronic acid

  • MDL number MFCD11976642
  • Cas Number : 860617-71-6
  • Molecular Weight : 149.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2195
Availability details for
Quantity
  • Availability
  • Add to Cart