COM448618200 Combi-Blocks, Inc.

4-(2-Chlorophenylmethoxy)phenylboronic acid

  • MDL number MFCD11540655
  • Cas Number : 871125-90-5
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2255
Availability details for
Quantity
  • Availability
  • Add to Cart