COM448618221 Combi-Blocks, Inc.

3-Formylpyridine-4-boronic acid

  • MDL number MFCD06409951
  • Cas Number : 1256355-58-4
  • Molecular Weight : 150.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2326
Availability details for
Quantity
  • Availability
  • Add to Cart