COM448618231 Combi-Blocks, Inc.

4-(N,N-Diethylaminomethyl)benzeneboronic acid

  • MDL number MFCD04039019
  • Cas Number : 220999-48-4
  • Molecular Weight : 207.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2358

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2358
Availability details for
Quantity
  • Availability
  • Add to Cart