COM448618232 Combi-Blocks, Inc.

3-Methoxythiophene-2-boronic acid

  • MDL number MFCD09952077
  • Cas Number : 162607-22-9
  • Molecular Weight : 158.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2369
Availability details for
Quantity
  • Availability
  • Add to Cart