COM448618239 Combi-Blocks, Inc.

6-Phenylpyridine-2-boronic acid

  • MDL number MFCD11223197
  • Cas Number : 1218790-96-5
  • Molecular Weight : 199.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2411
Availability details for
Quantity
  • Availability
  • Add to Cart