COM448618251 Combi-Blocks, Inc.

4-Chloro-1h-indole-2-boronic acid

  • MDL number MFCD11506061
  • Cas Number : 1256355-59-5
  • Molecular Weight : 195.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2500
Availability details for
Quantity
  • Availability
  • Add to Cart