COM448618253 Combi-Blocks, Inc.

4-Methoxyindole-2-boronic acid

  • MDL number MFCD11858386
  • Cas Number : 1256355-60-8
  • Molecular Weight : 191.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2508
Availability details for
Quantity
  • Availability
  • Add to Cart