COM448618257 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)pyridine-3-boronic acid

  • MDL number MFCD18258839
  • Cas Number : 1621416-45-2
  • Molecular Weight : 193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2643
Availability details for
Quantity
  • Availability
  • Add to Cart