COM448618261 Combi-Blocks, Inc.

2,3-Difluoropyridine-5-boronic acid

  • MDL number MFCD21609554
  • Cas Number : 1366482-40-7
  • Molecular Weight : 158.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2697

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2697
Availability details for
Quantity
  • Availability
  • Add to Cart