COM448618267 Combi-Blocks, Inc.

4-(4-Methyl-piperazinesulfonyl)phenyl boronic acid

  • MDL number MFCD12031677
  • Cas Number : 486422-11-1
  • Molecular Weight : 284.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(4-methyl-1-piperazinyl)sulfonyl]phenylboronic acid
Inchi Code 1S/C11H17BN2O4S/c1-13-6-8-14(9-7-13)19(17,18)11-4-2-10(3-5-11)12(15)16/h2-5,15-16H,6-9H2,1H3
Inchi Key WTIRVOVQNGLJIQ-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2795
Availability details for
Quantity
  • Availability
  • Add to Cart