COM448618277 Combi-Blocks, Inc.

5-(Methoxycarbonyl)furan-2-boronic acid

  • MDL number MFCD16660203
  • Cas Number : 876189-20-7
  • Molecular Weight : 169.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3158

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3158
Availability details for
Quantity
  • Availability
  • Add to Cart