COM448618281 Combi-Blocks, Inc.

2-(Methylsulfonyl)pyridine-5-boronic acid

  • MDL number MFCD12964553
  • Cas Number : 1088496-41-6
  • Molecular Weight : 201.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3276
Availability details for
Quantity
  • Availability
  • Add to Cart