COM448618289 Combi-Blocks, Inc.

1H-1,2,3-Benzotriazole-5-boronic acid

  • MDL number MFCD06254596
  • Cas Number : 183282-45-3
  • Molecular Weight : 162.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3744
Availability details for
Quantity
  • Availability
  • Add to Cart