COM448618314 Combi-Blocks, Inc.

5-Amino-2-chlorophenylboronic acid, HCl

  • MDL number MFCD12407139
  • Cas Number : 958646-69-0
  • Molecular Weight : 207.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3971
Availability details for
Quantity
  • Availability
  • Add to Cart