COM448618320 Combi-Blocks, Inc.

2-(3-Cyanophenylmethoxy)phenylboronic acid

  • MDL number MFCD12463462
  • Cas Number : 1256355-67-5
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4006
Availability details for
Quantity
  • Availability
  • Add to Cart