COM448618329 Combi-Blocks, Inc.

4-[(2,3-difluorophenyl)methoxy]phenylboronic acid

  • MDL number MFCD12498115
  • Cas Number : 1256355-76-6
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4025
Availability details for
Quantity
  • Availability
  • Add to Cart