COM448618334 Combi-Blocks, Inc.

2-(2,3-Difluorophenylmethoxy)phenyl)boronic acid

  • MDL number MFCD12591675
  • Cas Number : 1256355-81-3
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4037

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4037
Availability details for
Quantity
  • Availability
  • Add to Cart