COM448618335 Combi-Blocks, Inc.

2-(3-Fluorophenylmethoxy)-5-methylphenylboronic acid

  • MDL number MFCD12558059
  • Cas Number : 1256355-82-4
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4038
Availability details for
Quantity
  • Availability
  • Add to Cart