COM448618336 Combi-Blocks, Inc.

2-(3-Chlorophenylmethoxy)-5-fluorophenylboronic acid

  • MDL number MFCD12558063
  • Cas Number : 1256355-83-5
  • Molecular Weight : 280.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4041
Availability details for
Quantity
  • Availability
  • Add to Cart