COM448618337 Combi-Blocks, Inc.

2-(3,4-Dichlorophenylmethoxy)phenylboronic acid

  • MDL number MFCD12634040
  • Cas Number : 1256355-84-6
  • Molecular Weight : 296.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4045
Availability details for
Quantity
  • Availability
  • Add to Cart