COM448618343 Combi-Blocks, Inc.

3-(3,4-Dichlorophenylmethoxy)phenylboronic acid

  • MDL number MFCD12634049
  • Cas Number : 1256358-47-0
  • Molecular Weight : 296.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4053
Availability details for
Quantity
  • Availability
  • Add to Cart