COM448618351 Combi-Blocks, Inc.

4-(Methoxy-d3)phenylboronic acid

  • MDL number MFCD12760203
  • Cas Number : 1398503-83-7
  • Molecular Weight : 155.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4111
Availability details for
Quantity
  • Availability
  • Add to Cart