COM448618355 Combi-Blocks, Inc.

4-(3-methoxypropoxy)phenylboronic acid

  • MDL number MFCD12807145
  • Cas Number : 279262-35-0
  • Molecular Weight : 210.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4162
Availability details for
Quantity
  • Availability
  • Add to Cart