COM448618357 Combi-Blocks, Inc.

6-(Pyridin-3-yloxy)pyridine-3-boronic acid

  • MDL number MFCD12911765
  • Cas Number : 918138-38-2
  • Molecular Weight : 216.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4196
Availability details for
Quantity
  • Availability
  • Add to Cart