COM448618375 Combi-Blocks, Inc.

5-(2-Methoxyethoxy)pyridine-3-boronic acid

  • MDL number MFCD13183780
  • Cas Number : 1015229-31-8
  • Molecular Weight : 197.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4340
Availability details for
Quantity
  • Availability
  • Add to Cart