COM448618385 Combi-Blocks, Inc.

6-Ethoxy-5-(trifluoromethyl)-3-pyridinyl boronic acid

  • MDL number MFCD13195241
  • Cas Number : 849934-85-6
  • Molecular Weight : 235.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4452
Availability details for
Quantity
  • Availability
  • Add to Cart