COM448618386 Combi-Blocks, Inc.

2-Benzyloxy-5-(trifluoromethyl)pyridine-3-boronic acid

  • MDL number MFCD13195242
  • Cas Number : 850864-60-7
  • Molecular Weight : 297.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4453
Availability details for
Quantity
  • Availability
  • Add to Cart