COM448618388 Combi-Blocks, Inc.

2-(4-Trifluoromethylphenoxy)pyridine-5-boronic acid

  • MDL number MFCD14581929
  • Cas Number : 1256358-58-3
  • Molecular Weight : 283.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4474
Availability details for
Quantity
  • Availability
  • Add to Cart