COM448618389 Combi-Blocks, Inc.

2,3-Difluoro-4-propoxyphenylboronic acid

  • MDL number MFCD14584457
  • Cas Number : 212837-49-5
  • Molecular Weight : 216.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4493
Availability details for
Quantity
  • Availability
  • Add to Cart