COM448618390 Combi-Blocks, Inc.

3-(4-Fluorophenoxy)phenylboronic acid

  • MDL number MFCD14584726
  • Cas Number : 283173-82-0
  • Molecular Weight : 232.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4495
Availability details for
Quantity
  • Availability
  • Add to Cart