COM448618393 Combi-Blocks, Inc.

3-(Thiomorpholin-4-ylmethyl)phenylboronic acid

  • MDL number MFCD14687174
  • Cas Number : 1256358-59-4
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4536

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4536
Availability details for
Quantity
  • Availability
  • Add to Cart