COM448618400 Combi-Blocks, Inc.

4-(propoxymethyl)phenylboronic acid

  • MDL number MFCD14687314
  • Cas Number : 160061-48-3
  • Molecular Weight : 194.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4671
Availability details for
Quantity
  • Availability
  • Add to Cart