COM448618404 Combi-Blocks, Inc.

(1-Methyl-1H-benzimidazol-5-yl)boronic acid

  • MDL number MFCD16036273
  • Cas Number : 1107627-21-3
  • Molecular Weight : 176.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4841
Availability details for
Quantity
  • Availability
  • Add to Cart