COM448618407 Combi-Blocks, Inc.

2-(Cyclohexyloxy)methylphenylboronic acid

  • MDL number MFCD16198472
  • Cas Number : 498574-67-7
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5339
Availability details for
Quantity
  • Availability
  • Add to Cart