COM448618408 Combi-Blocks, Inc.

4-(Cyclohexyloxy)methylphenylboronic acid

  • MDL number MFCD16198473
  • Cas Number : 1221823-93-3
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5340
Availability details for
Quantity
  • Availability
  • Add to Cart