COM448618409 Combi-Blocks, Inc.

4-[(furan-2-ylmethoxy)methyl]phenylboronic acid

  • MDL number MFCD16198493
  • Cas Number : 1256358-66-3
  • Molecular Weight : 232.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5360
Availability details for
Quantity
  • Availability
  • Add to Cart