COM448618410 Combi-Blocks, Inc.

3-(2-Chlorophenoxymethyl)phenylboronic acid

  • MDL number MFCD16198497
  • Cas Number : 1256358-67-4
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5364
Availability details for
Quantity
  • Availability
  • Add to Cart