COM448618415 Combi-Blocks, Inc.

4-(4-chlorophenoxymethyl)phenylboronic acid

  • MDL number MFCD16198502
  • Cas Number : 1256358-72-1
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5369
Availability details for
Quantity
  • Availability
  • Add to Cart