COM448618418 Combi-Blocks, Inc.

4-(3-chlorophenoxymethyl)phenylboronic acid

  • MDL number MFCD16198518
  • Cas Number : 1256358-75-4
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5387
Availability details for
Quantity
  • Availability
  • Add to Cart