COM448618419 Combi-Blocks, Inc.

3-(Tetrahydropyran-4-yloxymethy)phenylboronic acid

  • MDL number MFCD16198628
  • Cas Number : 1256358-76-5
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5497
Availability details for
Quantity
  • Availability
  • Add to Cart