COM448618422 Combi-Blocks, Inc.

3-Fluoro-4-(thiophen-2-ylmethoxymethyl)phenylboronic acid

  • MDL number MFCD16198633
  • Cas Number : 1256358-79-8
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5502
Availability details for
Quantity
  • Availability
  • Add to Cart