COM448618423 Combi-Blocks, Inc.

3-(Thiophen-2-ylmethoxymethyl)phenylboronic acid

  • MDL number MFCD16198634
  • Cas Number : 1256358-80-1
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5503
Availability details for
Quantity
  • Availability
  • Add to Cart