COM448618433 Combi-Blocks, Inc.

2-(Hydroxymethyl)pyridine-4-boronic acid

  • MDL number MFCD22205764
  • Cas Number : 1414973-10-6
  • Molecular Weight : 152.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-5835
Availability details for
Quantity
  • Availability
  • Add to Cart