COM448618435 Combi-Blocks, Inc.

3-Bromo-5-iodophenylboronic acid

  • MDL number MFCD18383400
  • Cas Number : 1257793-09-1
  • Molecular Weight : 326.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6175

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6175
Availability details for
Quantity
  • Availability
  • Add to Cart