COM448618438 Combi-Blocks, Inc.

4-Acetylthiophene-2-boronic acid

  • MDL number MFCD18311854
  • Cas Number : 689247-77-6
  • Molecular Weight : 170.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6254
Availability details for
Quantity
  • Availability
  • Add to Cart