COM448618442 Combi-Blocks, Inc.

3-(N-BOC-Piperidin-4-ylmethoxy)phenylboronic acid

  • MDL number MFCD18783160
  • Cas Number : 1310404-86-4
  • Molecular Weight : 335.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6524
Availability details for
Quantity
  • Availability
  • Add to Cart