COM448618444 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-4-fluorophenylboronic acid

  • MDL number MFCD19981539
  • Cas Number : 1627839-42-2
  • Molecular Weight : 210.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6663
Availability details for
Quantity
  • Availability
  • Add to Cart