COM448618446 Combi-Blocks, Inc.

1-Cyclopentyl-1H-pyrazole-4-boronic acid

  • MDL number MFCD18250636
  • Cas Number : 1416786-06-5
  • Molecular Weight : 180

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-6671
Availability details for
Quantity
  • Availability
  • Add to Cart